Assay Details
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504855.2 | XP_003504903.2 | 2 - 3 | 256 | 83 | |
GenBank mRNA | JI882346.1 | - | 2 - 3 | 256 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
