Assay Details
| Gene Symbol: Efs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506133.2 | XP_003506181.1 | 3 - 4 | 1262 | 69 | |
| GenBank mRNA | JI891026.1 | - | 3 - 4 | 594 | 69 | |
| JP046110.1 | - | 3 - 4 | 594 | 69 | ||
More Information
Set Membership: |
|

Efs
Genomic Map