Assay Details
| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641070.1 | XP_007639260.1 | 2 - 3 | 394 | 103 | |
| GenBank mRNA | JI878130.1 | - | 2 - 3 | 193 | 103 | |
More Information
Set Membership: |
|

Cecr5
Genomic Map