Assay Details
Gene Symbol: Cecr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641070.1 | XP_007639260.1 | 4 - 5 | 518 | 64 | |
GenBank mRNA | JI878130.1 | - | 4 - 5 | 317 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
