Assay Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649300.1 | XP_007647490.1 | 6 - 7 | 711 | 61 | |
| GenBank mRNA | JP053011.1 | - | 1 | 246 | 61 | |
More Information
Set Membership: |
|

Dhdh
Genomic Map