Assay Details
| Gene Symbol: Agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653449.1 | XP_007651639.1 | 2 - 3 | 898 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agt| Gene Symbol: Agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653449.1 | XP_007651639.1 | 2 - 3 | 898 | 60 | |
Set Membership: |
|