Assay Details
| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502480.2 | XP_003502528.2 | 2 - 3 | 447 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata2| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502480.2 | XP_003502528.2 | 2 - 3 | 447 | 109 | |
Set Membership: |
|