Assay Details
| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507796.2 | XP_003507844.1 | 3 - 4 | 270 | 79 | |
| GenBank mRNA | JP053232.1 | - | 2 - 3 | 215 | 79 | |
More Information
Set Membership: |
|

Znrf2
Genomic Map