Assay Details
Gene Symbol: Qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507855.1 | XP_003507903.1 | 4 - 5 | 721 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
