Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648386.1 | XP_007646576.1 | 5 - 6 | 995 | 91 | |
| XM_007648387.1 | XP_007646577.1 | 5 - 6 | 941 | 91 | ||
| XM_007648388.1 | XP_007646578.1 | 5 - 6 | 929 | 91 | ||
| XM_007648389.1 | XP_007646579.1 | 5 - 6 | 928 | 91 | ||
More Information
Set Membership: |
|

Pvrl3
Genomic Map