Assay Details
| Gene Symbol: Ndufv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649025.1 | XP_007647215.1 | 7 - 8 | 710 | 63 | |
| XM_007649026.1 | XP_007647216.1 | 7 - 8 | 684 | 63 | ||
More Information
Set Membership: |
|

Ndufv2
Genomic Map