Assay Details
| Gene Symbol: Dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649182.1 | XP_007647372.1 | 4 - 5 | 609 | 61 | |
| XM_007649188.1 | XP_007647378.1 | 6 - 7 | 783 | 61 | ||
| XM_007649192.1 | XP_007647382.1 | 5 - 6 | 664 | 61 | ||
| GenBank mRNA | JP058419.1 | - | 1 - 2 | 249 | 61 | |
More Information
Set Membership: |
|

Dhrs7b
Genomic Map