Assay Details
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645053.1 | XP_007643243.1 | 6 - 7 | 749 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
