Assay Details
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501234.2 | XP_003501282.1 | 2 - 3 | 341 | 72 | |
XM_007641916.1 | XP_007640106.1 | 2 - 3 | 343 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
