Assay Details
Gene Symbol: Ldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508041.2 | XP_003508089.1 | 5 - 6 | 445 | 61 | |
XM_007648663.1 | XP_007646853.1 | 5 - 6 | 354 | 61 | ||
XM_007648664.1 | XP_007646854.1 | 5 - 6 | 354 | 61 | ||
GenBank mRNA | JI872359.1 | - | 4 - 5 | 257 | 61 | |
JP055663.1 | - | 4 - 5 | 257 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
