Assay Details
| Gene Symbol: Acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652527.1 | XP_007650717.1 | 4 - 5 | 292 | 77 | |
| GenBank mRNA | JI880976.1 | - | 3 - 4 | 166 | 78 | |
More Information
Set Membership: |
|

Acyp2
Genomic Map