Assay Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648092.1 | XP_007646282.1 | 2 - 3 | 299 | 91 | |
XM_007648093.1 | XP_007646283.1 | 3 - 4 | 303 | 91 | ||
XM_007648094.1 | XP_007646284.1 | 4 - 5 | 414 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
