Assay Details
| Gene Symbol: Cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497585.2 | XP_003497633.1 | 5 - 6 | 924 | 69 | |
| XM_007614324.1 | XP_007612514.1 | 6 - 7 | 967 | 69 | ||
| XM_007614339.1 | XP_007612529.1 | 6 - 7 | 968 | 69 | ||
| XM_007614347.1 | XP_007612537.1 | 5 - 6 | 925 | 69 | ||
| GenBank mRNA | JI893032.1 | - | 2 - 3 | 309 | 69 | |
| JP042047.1 | - | 2 - 3 | 231 | 69 | ||
| JP054663.1 | - | 2 - 3 | 307 | 69 | ||
More Information
Set Membership: |
|

Cnot7
Genomic Map