Assay Details
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510606.1 | XP_003510654.1 | 1 - 2 | 128 | 104 | |
XM_007651230.1 | XP_007649420.1 | 1 - 2 | 128 | 104 | ||
XM_007651231.1 | XP_007649421.1 | 1 - 2 | 128 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
