Assay Details
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513976.2 | XP_003514024.1 | 3 - 4 | 466 | 90 | |
XM_007654197.1 | XP_007652387.1 | 4 - 5 | 695 | 90 | ||
XM_007654198.1 | XP_007652388.1 | 3 - 4 | 559 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
