Assay Details
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007634128.1 | XP_007632318.1 | 1 - 2 | 121 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
