Assay Details
| Gene Symbol: Apex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500185.2 | XP_003500233.1 | 5 - 6 | 728 | 56 | |
| GenBank mRNA | JI892189.1 | - | 3 - 4 | 247 | 56 | |
| JP043864.1 | - | 2 - 3 | 216 | 56 | ||
More Information
Set Membership: |
|

Apex2
Genomic Map