Assay Details
| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502566.2 | XP_003502614.1 | 2 - 3 | 381 | 68 | |
| GenBank mRNA | JI873600.1 | - | 2 - 3 | 188 | 68 | |
| JP055668.1 | - | 2 - 3 | 188 | 68 | ||
More Information
Set Membership: |
|

Atp5d
Genomic Map