Assay Details
| Gene Symbol: Ndufa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505971.2 | XP_003506019.1 | 2 - 3 | 260 | 65 | |
| GenBank mRNA | JI894452.1 | - | 2 - 3 | 254 | 65 | |
More Information
Set Membership: |
|

Ndufa4
Genomic Map