Assay Details
| Gene Symbol: Wibg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506201.2 | XP_003506249.2 | 1 - 2 | 406 | 67 | |
| XM_007646705.1 | XP_007644895.1 | 1 - 2 | 415 | 67 | ||
More Information
Set Membership: |
|

Wibg
Genomic Map