Assay Details
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507229.2 | XP_003507277.1 | 6 - 7 | 983 | 59 | |
GenBank mRNA | JP056645.1 | - | 1 - 2 | 72 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
