Assay Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510738.2 | XP_003510786.1 | 3 - 4 | 406 | 67 | |
| GenBank mRNA | JI867834.1 | - | 2 - 3 | 368 | 67 | |
| JP049723.1 | - | 2 - 3 | 368 | 67 | ||
More Information
Set Membership: |
|

Arf3
Genomic Map