Assay Details
Gene Symbol: Mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510867.2 | XP_003510915.2 | 4 - 5 | 623 | 94 | |
GenBank mRNA | JI893925.1 | - | 4 - 5 | 547 | 94 | |
JP052734.1 | - | 4 - 5 | 547 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
