Assay Details
Gene Symbol: Ccl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500946.1 | XP_003500994.1 | 1 - 2 | 71 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
