Assay Details
| Gene Symbol: LOC100756511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501029.2 | XP_003501077.2 | 6 - 7 | 1500 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756511| Gene Symbol: LOC100756511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501029.2 | XP_003501077.2 | 6 - 7 | 1500 | 98 | |
Set Membership: |
|