Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506176.2 | XP_003506224.1 | 9 - 10 | 1526 | 91 | |
| GenBank mRNA | JI874525.1 | - | 8 - 9 | 1291 | 91 | |
| JP060300.1 | - | 8 - 9 | 1291 | 91 | ||
More Information
Set Membership: |
|

Atp5b
Genomic Map