Assay Details
| Gene Symbol: Shc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506251.2 | XP_003506299.1 | 3 - 4 | 1313 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shc4| Gene Symbol: Shc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506251.2 | XP_003506299.1 | 3 - 4 | 1313 | 58 | |
Set Membership: |
|