Assay Details
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506264.2 | XP_003506312.1 | 3 - 4 | 244 | 79 | |
GenBank mRNA | JI869439.1 | - | 1 - 2 | 57 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
