Assay Details
| Gene Symbol: Acd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508554.2 | XP_003508602.1 | 2 - 3 | 312 | 55 | |
| XM_007649181.1 | XP_007647371.1 | 2 - 3 | 312 | 55 | ||
| GenBank mRNA | JI863880.1 | - | 1 - 2 | 134 | 55 | |
| JP056583.1 | - | 1 - 2 | 134 | 55 | ||
More Information
Set Membership: |
|

Acd
Genomic Map