Assay Details
| Gene Symbol: Adora3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654508.1 | XP_007652698.1 | 5 - 6 | 615 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adora3| Gene Symbol: Adora3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654508.1 | XP_007652698.1 | 5 - 6 | 615 | 109 | |
Set Membership: |
|