Assay Details
Gene Symbol: Syde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645208.1 | XP_007643398.1 | 6 - 7 | 1854 | 56 | |
XM_007645209.1 | XP_007643399.1 | 6 - 7 | 1818 | 56 | ||
GenBank mRNA | JI862517.1 | - | 1 - 2 | 160 | 56 | |
JP063660.1 | - | 1 - 2 | 160 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
