Assay Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654283.1 | XP_007652473.1 | 2 - 3 | 187 | 62 | |
XM_007654284.1 | XP_007652474.1 | 2 - 3 | 185 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
