Assay Details
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498937.2 | XP_003498985.1 | 2 - 3 | 558 | 67 | |
XM_007635439.1 | XP_007633629.1 | 2 - 3 | 768 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
