Assay Details
Gene Symbol: Cdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506838.2 | XP_003506886.2 | 5 - 6 | 722 | 62 | |
XM_007647367.1 | XP_007645557.1 | 5 - 6 | 654 | 62 | ||
XM_007647368.1 | XP_007645558.1 | 6 - 7 | 617 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
