Assay Details
| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI869661.1 | - | 3 - 4 | 532 | 77 | |
| JP055313.1 | - | 3 - 4 | 532 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763249| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI869661.1 | - | 3 - 4 | 532 | 77 | |
| JP055313.1 | - | 3 - 4 | 532 | 77 | ||