Assay Details
Gene Symbol: Spint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645130.1 | XP_007643320.1 | 1 - 2 | 111 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
