Assay Details
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500901.2 | XP_003500949.1 | 3 - 4 | 623 | 71 | |
GenBank mRNA | JI864489.1 | - | 2 - 3 | 346 | 71 | |
JP044218.1 | - | 2 - 3 | 346 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
