Assay Details
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507942.2 | XP_003507990.1 | 2 - 3 | 298 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
