Assay Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501605.2 | XP_003501653.1 | 2 - 3 | 440 | 58 | |
GenBank mRNA | JI889691.1 | - | 2 - 3 | 429 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
