Assay Details
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502509.2 | XP_003502557.1 | 1 - 2 | 243 | 92 | |
GenBank mRNA | JP064227.1 | - | 1 - 2 | 55 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
