Assay Details
| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507904.2 | XP_003507952.1 | 7 - 8 | 1159 | 61 | |
| GenBank mRNA | JI894445.1 | - | 7 - 8 | 828 | 61 | |
| JP048511.1 | - | 7 - 8 | 827 | 61 | ||
More Information
Set Membership: |
|

Imp4
Genomic Map