Assay Details
| Gene Symbol: Swi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652779.1 | XP_007650969.1 | 1 - 2 | 155 | 79 | |
| GenBank mRNA | JP047327.1 | - | 335 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Swi5| Gene Symbol: Swi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652779.1 | XP_007650969.1 | 1 - 2 | 155 | 79 | |
| GenBank mRNA | JP047327.1 | - | 335 | 79 | ||
Set Membership: |
|