Assay Details
Gene Symbol: Cmtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499685.2 | XP_003499733.1 | 4 - 5 | 446 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
