Assay Details
Gene Symbol: Mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506801.1 | XP_003506849.1 | 1 - 2 | 542 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
