Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513708.2 | XP_003513756.2 | 3 - 4 | 1069 | 54 | |
| GenBank mRNA | JI872076.1 | - | 1 - 2 | 178 | 54 | |
| JP064921.1 | - | 3 - 4 | 844 | 54 | ||
More Information
Set Membership: |
|

Anapc2
Genomic Map